COM448686385 Combi-Blocks, Inc.

Methyl (2r)-2-amino-3-(4-cyanophenyl)propanoate

  • MDL number MFCD09255818
  • Cas Number : 190060-97-0
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1099
Availability details for
Quantity
  • Availability
  • Add to Cart