COM448686387 Combi-Blocks, Inc.

3-Hydroxy-3-phenyl-propionic acid methyl ester

  • MDL number MFCD19691249
  • Cas Number : 7497-61-2
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1102
Availability details for
Quantity
  • Availability
  • Add to Cart