COM448686388 Combi-Blocks, Inc.

Methyl 2-([(tert-butoxy)carbonyl](ethyl)amino)acetate

  • MDL number MFCD16053518
  • Cas Number : 1249016-05-4
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1104

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1104
Availability details for
Quantity
  • Availability
  • Add to Cart