COM448686391 Combi-Blocks, Inc.

Methyl 5,6,7,8-tetrahydronaphthalene-1-carboxylate

  • MDL number MFCD15523539
  • Cas Number : 66193-59-7
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1111
Availability details for
Quantity
  • Availability
  • Add to Cart