COM448686392 Combi-Blocks, Inc.

methyl 2-[(2S)-2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanamido]acetate

  • MDL number MFCD27578352
  • Cas Number : 27610-07-7
  • Molecular Weight : 302.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1112

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1112
Availability details for
Quantity
  • Availability
  • Add to Cart