COM448686397 Combi-Blocks, Inc.

methyl 2-[(2-methylpropyl)sulfanyl]benzoate

  • MDL number MFCD27578354
  • Cas Number : 1794367-72-8
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1119
Availability details for
Quantity
  • Availability
  • Add to Cart