COM448686398 Combi-Blocks, Inc.

methyl (2S)-2-{[(benzyloxy)carbonyl](methyl)amino}propanoate

  • MDL number MFCD27578355
  • Cas Number : 24164-72-5
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1120

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1120
Availability details for
Quantity
  • Availability
  • Add to Cart