COM448686402 Combi-Blocks, Inc.

2-Methyl-1h-indole-3-acetic acid methyl ester

  • MDL number MFCD11486692
  • Cas Number : 78564-10-0
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1128

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1128
Availability details for
Quantity
  • Availability
  • Add to Cart