COM448686408 Combi-Blocks, Inc.

Methyl (2r)-2,3-dihydro-1h-indole-2-carboxylate

  • MDL number MFCD16037973
  • Cas Number : 293737-30-1
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1136
Availability details for
Quantity
  • Availability
  • Add to Cart