COM448686413 Combi-Blocks, Inc.

Methyl 5-(3-chlorophenyl)-1h-pyrazole-3-carboxylate

  • MDL number MFCD12188758
  • Cas Number : 1187360-62-8
  • Molecular Weight : 236.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1143

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1143
Availability details for
Quantity
  • Availability
  • Add to Cart