COM448686415 Combi-Blocks, Inc.

Namiki bbv-45525676

  • MDL number MFCD23720399
  • Cas Number : 1014631-07-2
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1146

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1146
Availability details for
Quantity
  • Availability
  • Add to Cart