COM448686417 Combi-Blocks, Inc.

methyl 2-[2-bromo-4-(trifluoromethyl)phenyl]acetate

  • MDL number MFCD27578361
  • Cas Number : 1069115-04-3
  • Molecular Weight : 297.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1152

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1152
Availability details for
Quantity
  • Availability
  • Add to Cart