COM448686419 Combi-Blocks, Inc.

Methyl 4-chloro-3-(propan-2-yloxy)benzoate

  • MDL number MFCD19440114
  • Cas Number : 1394933-47-1
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1154

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1154
Availability details for
Quantity
  • Availability
  • Add to Cart