COM448686421 Combi-Blocks, Inc.

(S)-2-Amino-pent-4-enoic acid methyl ester

  • MDL number MFCD19203463
  • Cas Number : 50299-15-5
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1160

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1160
Availability details for
Quantity
  • Availability
  • Add to Cart