COM448686423 Combi-Blocks, Inc.

methyl (2R)-3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate

  • MDL number MFCD27578363
  • Cas Number : 1820580-35-5
  • Molecular Weight : 397.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1163
Availability details for
Quantity
  • Availability
  • Add to Cart