COM448686427 Combi-Blocks, Inc.

methyl 2-[5-fluoro-2-(trifluoromethyl)phenyl]acetate

  • MDL number MFCD27578364
  • Cas Number : 1820703-52-3
  • Molecular Weight : 236.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1170

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1170
Availability details for
Quantity
  • Availability
  • Add to Cart