COM448686428 Combi-Blocks, Inc.

Methyl 4-chloro-3-propoxybenzoate

  • MDL number MFCD19440098
  • Cas Number : 69076-29-5
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1173

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1173
Availability details for
Quantity
  • Availability
  • Add to Cart