COM448686431 Combi-Blocks, Inc.

(R)-Methyl 5-oxo-1-((R)-1-phenylethyl)pyrrolidine-3-carboxylate

  • MDL number MFCD12963552
  • Cas Number : 852857-11-5
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1184

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1184
Availability details for
Quantity
  • Availability
  • Add to Cart