COM448686435 Combi-Blocks, Inc.

methyl 2-(2,5-dibromophenyl)acetate

  • MDL number MFCD27578366
  • Cas Number : 203314-32-3
  • Molecular Weight : 308.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1192

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1192
Availability details for
Quantity
  • Availability
  • Add to Cart