COM448686436 Combi-Blocks, Inc.

methyl 4-bromo-2-butoxybenzoate

  • MDL number MFCD27578367
  • Cas Number : 1820640-94-5
  • Molecular Weight : 287.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1196

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1196
Availability details for
Quantity
  • Availability
  • Add to Cart