COM448686453 Combi-Blocks, Inc.

Methyl (2r)-2-[(tert-butoxy)carbonylamino]-3-(4-fluorophenyl)propanoate

  • MDL number MFCD14636381
  • Cas Number : 176794-99-3
  • Molecular Weight : 297.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1228

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1228
Availability details for
Quantity
  • Availability
  • Add to Cart