COM448686454 Combi-Blocks, Inc.

Methyl 2,5-dimethyloxazole-4-carboxylate

  • MDL number MFCD23159800
  • Cas Number : 73537-07-2
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1229

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1229
Availability details for
Quantity
  • Availability
  • Add to Cart