COM448686455 Combi-Blocks, Inc.

(S)-Methyl 5-oxo-1-((S)-1-phenylethyl)pyrrolidine-3-carboxylate

  • MDL number MFCD14635886
  • Cas Number : 99735-45-2
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1230

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1230
Availability details for
Quantity
  • Availability
  • Add to Cart