COM448686456 Combi-Blocks, Inc.

tert-butyl 3-[(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-methoxy-3-oxopropyl]indole-1-carboxylate

  • MDL number MFCD27578370
  • Cas Number : 405555-31-9
  • Molecular Weight : 540.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1231

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1231
Availability details for
Quantity
  • Availability
  • Add to Cart