COM448686461 Combi-Blocks, Inc.

6-Bromo-4-hydroxy-quinoline-3-carboxylic acid methyl ester

  • MDL number MFCD08436143
  • Cas Number : 1593717-84-0
  • Molecular Weight : 282.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1242

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1242
Availability details for
Quantity
  • Availability
  • Add to Cart