COM448686466 Combi-Blocks, Inc.

methyl (2E)-3-(3-bromo-2-fluorophenyl)prop-2-enoate

  • MDL number MFCD27578373
  • Cas Number : 1363403-01-3
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1259

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1259
Availability details for
Quantity
  • Availability
  • Add to Cart