COM448686469 Combi-Blocks, Inc.

[3-(methoxycarbonyl)-4-(trifluoromethoxy)phenyl]boronic acid

  • MDL number MFCD27578374
  • Molecular Weight : 264.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1268

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1268
Availability details for
Quantity
  • Availability
  • Add to Cart