COM448686471 Combi-Blocks, Inc.

methyl 4-bromo-2-(2-methylpropoxy)benzoate

  • MDL number MFCD27578375
  • Cas Number : 855482-60-9
  • Molecular Weight : 287.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1270
Availability details for
Quantity
  • Availability
  • Add to Cart