COM448686473 Combi-Blocks, Inc.

Cyclopropanecarboxylic acid, 1-(4-methoxyphenyl)-, methyl ester

  • MDL number MFCD20040021
  • Cas Number : 779206-51-8
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1273

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1273
Availability details for
Quantity
  • Availability
  • Add to Cart