COM448686475 Combi-Blocks, Inc.

Methyl 1-(4-chlorophenyl)cyclopentanecarboxylate

  • MDL number MFCD21333130
  • Cas Number : 914106-85-7
  • Molecular Weight : 238.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1275

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1275
Availability details for
Quantity
  • Availability
  • Add to Cart