COM448686478 Combi-Blocks, Inc.

Methyl 2-[(2-nitrophenyl)amino]propanoate

  • MDL number MFCD01364788
  • Cas Number : 1396964-74-1
  • Molecular Weight : 224.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1283

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1283
Availability details for
Quantity
  • Availability
  • Add to Cart