COM448686479 Combi-Blocks, Inc.

Methyl 3-(cyclopropylsulfamoyl)benzoate

  • MDL number MFCD15724900
  • Cas Number : 1456057-17-2
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1285

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1285
Availability details for
Quantity
  • Availability
  • Add to Cart