COM448686483 Combi-Blocks, Inc.

4-benzyl 1-methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578377
  • Cas Number : 80963-12-8
  • Molecular Weight : 337.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1294

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1294
Availability details for
Quantity
  • Availability
  • Add to Cart