COM448686486 Combi-Blocks, Inc.

methyl 3-ethoxy-2-fluorobenzoate

  • MDL number MFCD27578379
  • Cas Number : 1820604-99-6
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1301

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1301
Availability details for
Quantity
  • Availability
  • Add to Cart