COM448686487 Combi-Blocks, Inc.

methyl 4-butoxy-3-methoxybenzoate

  • MDL number MFCD09731411
  • Cas Number : 3535-28-2
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1302
Availability details for
Quantity
  • Availability
  • Add to Cart