COM448686493 Combi-Blocks, Inc.

methyl (2S)-2-phenylpropanoate

  • MDL number MFCD27578380
  • Cas Number : 28645-07-0
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1313

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1313
Availability details for
Quantity
  • Availability
  • Add to Cart