COM448686499 Combi-Blocks, Inc.

Methyl (3s)-3-boc-amino-3-phenylpropionate

  • MDL number MFCD16659973
  • Cas Number : 190189-97-0
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1328

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1328
Availability details for
Quantity
  • Availability
  • Add to Cart