COM448686500 Combi-Blocks, Inc.

Methyl 2-(4-methylcyclohexylidene)acetate

  • MDL number MFCD23724325
  • Cas Number : 54263-85-3
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1331

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1331
Availability details for
Quantity
  • Availability
  • Add to Cart