COM448686504 Combi-Blocks, Inc.

Methyl 3-(piperidine-1-sulfonyl)benzoate

  • MDL number MFCD00814101
  • Cas Number : 415712-10-6
  • Molecular Weight : 283.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1335
Availability details for
Quantity
  • Availability
  • Add to Cart