COM448686508 Combi-Blocks, Inc.

Methyl 5-bromo-2-hydroxy-3-methylbenzoate

  • MDL number MFCD16293433
  • Cas Number : 40912-71-8
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1341

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1341
Availability details for
Quantity
  • Availability
  • Add to Cart