COM448686509 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl](methyl)amino}-4-methylpentanoate

  • MDL number MFCD27578383
  • Cas Number : 53363-90-9
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1342

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1342
Availability details for
Quantity
  • Availability
  • Add to Cart