COM448686510 Combi-Blocks, Inc.

Methyl (2s)-2-amino-2-cyclopropylacetate

  • MDL number MFCD06200656
  • Cas Number : 693213-58-0
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1346

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1346
Availability details for
Quantity
  • Availability
  • Add to Cart