COM448686513 Combi-Blocks, Inc.

Methyl (3-oxo-2,3-dihydro-4h-1,4-benzoxazin-4-yl)acetate

  • MDL number MFCD04207474
  • Cas Number : 450358-64-2
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1350

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1350
Availability details for
Quantity
  • Availability
  • Add to Cart