COM448686517 Combi-Blocks, Inc.

Methyl benzo[d]thiazole-2-carboxylate

  • MDL number MFCD00848403
  • Cas Number : 87802-07-1
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1359

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1359
Availability details for
Quantity
  • Availability
  • Add to Cart