COM448686518 Combi-Blocks, Inc.

5,6-Difluoro-2-indole carboxylic acid methyl ester

  • MDL number MFCD08689964
  • Cas Number : 898746-30-0
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1360
Availability details for
Quantity
  • Availability
  • Add to Cart