COM448686519 Combi-Blocks, Inc.

Methyl 3-(propylsulfamoyl)benzoate

  • MDL number MFCD11618376
  • Cas Number : 1094563-62-8
  • Molecular Weight : 257.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1362
Availability details for
Quantity
  • Availability
  • Add to Cart