COM448686522 Combi-Blocks, Inc.

Cyclopentanecarboxylic acid, 1-[[(1,1-dimethylethoxy)carbonyl]amino]-, methyl ester

  • MDL number MFCD24392969
  • Cas Number : 248262-96-6
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1368
Availability details for
Quantity
  • Availability
  • Add to Cart