COM448686525 Combi-Blocks, Inc.

Methyl 3-(morpholine-4-sulfonyl)benzoate

  • MDL number MFCD01141529
  • Cas Number : 412940-98-8
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1371

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1371
Availability details for
Quantity
  • Availability
  • Add to Cart