COM448686526 Combi-Blocks, Inc.

methyl 3-{2-[2-(benzyloxy)ethoxy]ethoxy}propanoate

  • MDL number MFCD27578386
  • Cas Number : 127457-61-8
  • Molecular Weight : 282.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1372

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1372
Availability details for
Quantity
  • Availability
  • Add to Cart