COM448686528 Combi-Blocks, Inc.

Methyl 2-ethylbenzoate

  • MDL number MFCD06203917
  • Cas Number : 50604-01-8
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1374
Availability details for
Quantity
  • Availability
  • Add to Cart